missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite, 4-6%

Catalog No. p-3735121
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419550250 25 mL Plastic bottle
AC419552500 250 mL Plastic bottle
AC419550010 1 L Plastic bottle
AC419550025 2.5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC419550250 Supplier Thermo Scientific Chemicals Supplier No. 419550250
Only null left

CAS: 7681-52-9 | ClNaO | 74.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Color Yellow
Physical Form Solution
Quantity 25 mL
Linear Formula NaOCl
Fieser 01,1084; 04,456; 05,617; 06,543; 07,337; 08,461; 09,430; 10,365; 11,487; 15,293; 16,308
Merck Index 15,8762
Solubility Information Solubility in water: soluble
Formula Weight 74.44
Percent Purity 4.5 to 5.5%
Grade Reagent
Packaging Plastic bottle
Specific Gravity 1.083
Density 1.0830g/mL
Chemical Name or Material Sodium hypochlorite, 4-6% Chlorine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting effects.
May be corrosive to metals.
Contact with acids liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 231-668-3

RUO – Research Use Only