missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite, 4-6%

CAS: 7681-52-9 | ClNaO | 74.44 g/mol

Supplier:  Thermo Scientific Chemicals 419550010

 View more versions of this product

Catalog No. AC419550010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Sodium hypochlorite
7681-52-9,7732-18-5
Solution
ClNaO
MFCD00011120
15,8762
Solubility in water: soluble
[O-]Cl.[Na+]
74.44
CHEBI:32146
4.5 to 5.5%
Plastic bottle
1.0830g/mL
4-6% Chlorine
Yellow
1 L
NaOCl
01,1084; 04,456; 05,617; 06,543; 07,337; 08,461; 09,430; 10,365; 11,487; 15,293; 16,308
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
SUKJFIGYRHOWBL-UHFFFAOYSA-N
sodium;hypochlorite
23665760
74.44
Reagent
1.083

Chemical Identifiers

7681-52-9
74.44
SUKJFIGYRHOWBL-UHFFFAOYSA-N
23665760
sodium;hypochlorite
ClNaO
MFCD00011120
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
CHEBI:32146
[O-]Cl.[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting effects.
May be corrosive to metals.
Contact with acids liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 231-668-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only