missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide (White Pellets), Fisher BioReagents™

Catalog No. p-2363422
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP359500 500 g Poly Bottle
BP359212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP359500 Supplier Fisher BioReagents Supplier No. BP359 500
Only null left

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium Hydroxide (White Pellets)

Sodium Hydroxide is used to adjust the pH of buffers for molecular biology and cell culture applications.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
UN Number 1823
Quantity 500 g
Formula Weight 40
Percent Purity ≥97.0%
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 1mbar at 700°C
Physical Form Solid
Assay ≥98%
Chemical Name or Material Sodium Hydroxide
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Recommended Storage RT