missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide-Thiosulfate, 50%-2.5% (w/v), Ricca Chemical

Catalog No. 749516
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
749516 500 mL Natural Poly Bottle
749532 1 L Natural Poly Bottle
74951 4 L Natural Poly Bottle
749525 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 749516 Supplier Ricca Chemical Company Supplier No. 749516

For Analysis of Nitrogen (Kjeldahl) Using Copper Catalyst

Chemical Identifiers

Molecular Formula HNaO
Molecular Weight (g/mol) Mixture
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Min % 1.21, 48.29
Color Colorless
CAS Max % 1.26, 50.26
CAS 10102-17-7, 1310-73-2, 7732-18-5
Quantity 500 mL
Packaging Natural Poly Bottle
Grade Laboratory
Concentration or Composition (by Analyte or Components) 50% (Sodium Hydroxide) -2.5% (Thiosulfate) (w/v)
Chemical Name or Material Sodium Hydroxide-Thiosulfate Solution