missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N), Honeywell Fluka™

Volumetric, 0.5 M NaOH (0.5N)

$54.60 - $119.95

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6001440
View Documents
Honeywell-Fluka
319503500ML
500 mL Poly Bottle
Each for $54.60
Only null left
 
6001439
View Documents
Honeywell-Fluka
3195032L
2 L Poly Bottle
Each for $119.95
Only null left
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]

Specifications

>100°C
≥90.0000%,≥1.0000%
HNaO
MFCD00003548
UN1824
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40.00
Poly Bottle
1310-73-2,7732-18-5
≤100.0000%,<2.0000%
NaOH
0.5 M NaOH (0.5N)
1.02 g/mL at 25 °C
500 mL
[OH-].[Na+]
39.997
CHEBI:32145
Volumetric
Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N)
Videos
Safety and Handling

Safety and Handling

ShelfLife : 1080 days from date of manufacture

DOTInformation : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Sodium hydroxide solution

Recommended Storage : Room Temp

SDS
missing translation for 'documents'

missing translation for 'documents'