missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution (50% w/w/Certified), Fisher Chemical™

Supplier:  Fisher Chemical SS2544

 View more versions of this product

Catalog No. SS254-4

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

12.0
50% (w/w)
1310-73-2,7732-18-5
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Inclusive between 50%
Pass Test
3mmHg at 37°C
Sodium Hydroxide Solution
141.7°C
5°C
MFCD00003548
4 L
[OH-].[Na+]
39.997
CHEBI:32145
Certified
Poly Bottle
Liquid

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes severe burns by all exposure routes. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Accessories
Promotions

Promotions