missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, ≥32%, for determination of nitrogen, ≤0.0001% N, Honeywell™
≥32%, for determination of nitrogen, ≤0.0001% N
$173.70 - $2,378.58
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| MDL Number | MFCD00003548 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6001472
|
Honeywell Chemicals
305311L |
1 L | Poly Bottle |
N/A
|
|
||||
|
6001473
|
Honeywell Chemicals
3053125L |
2.5 L | Poly Bottle |
N/A
|
|
||||
|
6001474
|
Honeywell Chemicals
305315L |
5 L | Poly Bottle |
N/A
|
|
||||
|
60045830
|
Honeywell Chemicals
305316X1L |
6 x 1 L | Poly Bottle |
N/A
|
|
||||
|
60045828
|
Honeywell Chemicals
305314X25L |
4 x 2.5 L | Poly Bottle |
N/A
|
|
||||
|
60045829
|
Honeywell Chemicals
305314X5L |
4 x 5 L | Poly Bottle |
N/A
|
|
||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
Specifications
| 120°C | |
| ≥50.0000%,≥25.0000% | |
| HNaO | |
| MFCD00003548 | |
| 1.350 g/cm3 | |
| 1 L | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| >32% | |
| Sodium hydroxide solution, ≥32%, for determination of nitrogen, ≤0.0001% N |
| 1310-73-2,7732-18-5 | |
| <75.0000%,<50.0000% | |
| NaOH | |
| UN1824 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40.0 | |
| Poly Bottle |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
DOTInformation : Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sodium hydroxide solution
Recommended Storage : Room Temp