missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, 30.0%, Suprapur™, MilliporeSigma™
$608.62 - $608.62
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 40.00 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| IUPAC Name | sodium hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M1055890250
|
Technidata
1055890250 |
250 mL |
Each for $608.62
|
|
|||||
Safety and Handling
Recommended Storage : Store at 15 to 25°C