missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, 30.0%, Suprapur™, MilliporeSigma™

Catalog No. M1055890250
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1055890250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1055890250 Supplier Technidata Supplier No. 1.05589.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14 (H2O, 20°C)
Assay Percent Range ≥ 30.0%
Density 1.33 g/cm3 (at 20°C)
Quantity 250 mL
Packaging Plastic Bottle
Recommended Storage Store at 15 to 25°C