missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Honeywell Fluka™

Catalog No. 6001467
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001467 1 L
6001468 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001467 Supplier Honeywell Chemicals Supplier No. 716861L
Only null left

for HPLC, 50%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
  • Approx. molarity: 19M

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Assay Percent Range 49.0 - 51.0%
Linear Formula NaOH
Concentration 50% (w/w)
UN Number UN1824
Density 1.515g/mL at 20°C
Quantity 1 L
Explosion Limit for HPLC
Formula Weight 40g/mol
Percent Purity 50%
Grade HPLC
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide solution
Show More Show Less
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338