missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, 0.05M, Honeywell Fluka™
Volumetric, 0.05Â M NaOH (0.05N)
$169.21 - $1161.13
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| MDL Number | MFCD00003548 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6002624
|
Honeywell Chemicals
35249500ML |
500 mL |
Each for $169.21
|
|
|||||
|
6002623
|
Honeywell Chemicals
352491L |
1 L |
Each for $222.63
|
|
|||||
|
60046549
|
Honeywell Chemicals
352496X500ML |
6 x 500 mL |
Case of 6 Each for $873.97
|
|
|||||
|
60046548
|
Honeywell Chemicals
352496X1L |
6 x 1 L |
Case of 6 Each for $1,161.13
|
|
|||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
Specifications
| 1310-73-2 | |
| NaOH | |
| 0.05M | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40g/mol | |
| Sodium hydroxide solution |
| HNaO | |
| MFCD00003548 | |
| NONH for all modes of transport | |
| 500 mL | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| 3 mmHg (37°C) |