missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Semiconductor grade, Honeywell
Pellets, semiconductor grade, 99.99% trace metals basis
$454.25 - $1684.70
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| MDL Number | MFCD00003548 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
Description
- Comprehensive portfolio for the electronics and semiconductor industry
- For scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
- Superior purity, stability and exceptional lot-to-lot consistency
- Includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
Specifications
| 1310-73-2 | |
| NaOH | |
| 1823 | |
| Soluble in water, 1260 g/L at 20°C | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40g/mol | |
| Semiconductor | |
| 18mmHg (20°C), 3mmHg (37°C) |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| 100 g | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| 99.99% (trace metals basis) | |
| >1 (vs air) | |
| Sodium Hydroxide |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314
EINECSNumber : 215-185-5
RTECSNumber : WB4900000