missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Reagent Grade, 97%, 20 to 40 Mesh, Honeywell™

Reagent Grade, beads, 20-40 mesh, 97%

$397.18 - $397.18

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046650
View Documents
Honeywell Chemicals
36717625KG
2.5 kg Poly Bottle
Each for $397.18
Only null left
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]

Specifications

Colorless or White
1310-73-2
HNaO
MFCD00003548
2.13g/cm3
2.5 kg
[OH-].[Na+]
39.997
CHEBI:32145
97%
Poly Bottle
18mmHg (20°C), 3mmHg (37°C)
Sodium hydroxide
1390°C
318°C
NaOH
UN1823
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40g/mol
Reagent
>1 (vs air)
Beads
Videos
Safety and Handling

Safety and Handling

P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

DOTInformation : 8/PGII

RTECSNumber : 215-185-5

SDS
missing translation for 'documents'

missing translation for 'documents'