missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., K ≤0.02%, ≥98%, pellets, Honeywell Fluka™

Numéro de catalogue. 60045836
Change view
Click to view available options
Quantity:
4 x 5 kg
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
60045836 6 x 1 kg
60045835 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 60045836 Fournisseur Honeywell Chemicals Code fournisseur 306206X1KG
Il en reste null

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., K ≤0.02%,≥98%, pellets

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Identifiants chimiques

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Spécifications

Boiling Point 1,390°C
Melting Point 318°C (literature)
CAS Max % 100.0000%
Linear Formula NaOH
UN Number UN1823
Density 2.13 g/cm3
Quantity 6 x 1 kg
Formula Weight 40.00
Percent Purity ≥98%
Grade ACS Reagent/Reagent Ph. Eur.
Packaging Poly Bottle
Physical Form Pellet
Chemical Name or Material Sodium hydroxide, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Afficher plus de résultats Afficher moins de résultats
missing translation for 'shelfLife' 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sodium hydroxide, solid
EINECSNumber 215-185-5
Recommended Storage Room Temp