missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, puriss p.a., ≥98% (T), ACS Reagent, Honeywell Fluka™

Catalog No. 6001420
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001420 5 kg
6001419 500 g
6001418 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001420 Supplier Honeywell Chemicals Supplier No. 716905KG
Only null left

Puriss. p.a., ACS Reagent, K ≤0.02%,≥98.0% (T), pellets

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number UN1823
Quantity 5 kg
Formula Weight 40g/mol
Percent Purity ≥98% (T)
Vapor Density >1 (vs air)
Vapor Pressure 18 mmHg (20°C), 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
RTECSNumber WB4900000