missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, Puriss., meets analytical spec. of BP, NF, Ph. Eur., E524, 98-100.5%, pellets, Honeywell Fluka™

Catalog No. p-7071728
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001477 1 kg
6001478 5 kg
60044580 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001477 Supplier Honeywell Chemicals Supplier No. 062031KG
Only null left

Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, 98-100.5%, pellets

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number 1823
Solubility Information Soluble in water, 1260g/L at 20°C
Quantity 1 kg
Formula Weight 40g/mol
Percent Purity 98 to 100.5%
Grade puriss.
Vapor Density >1 (vs air)
Vapor Pressure 3mmHg (37°C), 18mmHg (20°C)
Chemical Name or Material Sodium Hydroxide, Meets analytical specification of Ph. Eur.BP, NF
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H314
EINECSNumber 215-185-5
RTECSNumber WB4900000