missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Pellets, Technical, Spectrum™ Chemical

Catalog No. 18613657
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613657 50 kg Fiber Drum
18613654 500 g Poly Bottle
18613655 2.5 kg Poly Pail
18613656 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613657 Supplier Spectrum Chemical Mfg Cor Supplier No. S123850KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1310-73-2 | 40.0 g/mol

Chemical Identifiers

CAS 1310-73-2

Specifications

CAS Max % 1
Quantity 50 kg
Molecular Weight (g/mol) 40.0
Grade Technical
Packaging Fiber Drum
Chemical Name or Material Sodium Hydroxide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.