missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Pellets, Technical, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 500 g |
| Grade | Technical |
| Packaging | Poly Bottle |
| Chemical Name or Material | Sodium Hydroxide |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.