missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide (Pellets/NF/FCC), Fisher Chemical™
Specifications
Specifications
| pH | 14 |
| Color | White |
| Boiling Point | 1390°C |
| Melting Point | 318°C |
| UN Number | 1823 |
| Density | 2.13g/cm³ |
| Quantity | 50 kg |
| Formula Weight | 40 |
| Percent Purity | 95.0 to 100.5% (NF) |
| Grade | FCC/NF |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Causes severe burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting. NFPA Health:3 Flammability:0 Instability:1 |
| DOTInformation | DOT Class 8, : Corrosive |