Learn More
Sodium Hydroxide (Pellets/NF/FCC), Fisher Chemical™
Supplier: Thermo Fisher Scientific S3203
Specifications
| 14 | |
| White | |
| 1310-73-2,497-19-8 | |
| HNaO | |
| 1823 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40 | |
| FCC/NF | |
| Poly Bottle | |
| 1mmHg at 739°C |
| Sodium Hydroxide | |
| 1390°C | |
| 318°C | |
| MFCD00003548 | |
| 2.13g/cm³ | |
| 3 kg | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| 95.0 to 100.5% (FCC) | |
| Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF) | |
| Meets Requirements (NF) | |
| Solid |
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
Safety and Handling
DANGER!
Emergency Overview
Causes severe burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:0
Instability:1
DOTInformation : DOT Class 8, : Corrosive