missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Pellets, NF, 95-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Hydroxide, Pellets, NF is used as a pH adjusting agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Percent Purity | 95 to 100.5% |
| Grade | NF |
| Packaging | Poly Bottle |
| Chemical Name or Material | Sodium Hydroxide |