missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Pellets (Max. 0.02% K), For Analysis, EMSURE™, MilliporeSigma™

Catalog No. p-7082921
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1064691000 1 kg Plastic Bottle
M1064695000 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1064691000 Supplier Technidata Supplier No. 1.06469.1000
Only null left

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH <14 (100g/L, H2O, 20°C)
Color White
Melting Point 319°C to 322°C
Density 2.13 g/3 (20 °C)
Solubility Information 1090g/L
Quantity 1 kg
Formula Weight 40
Percent Purity 99 to 100%
Grade ACS
Packaging Plastic Bottle
Physical Form Solid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
EINECSNumber 215-185-5