missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Pellets, FCC, 95-100.5%, Spectrum™ Chemical

Catalog No. 18606574
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606574 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606574 Supplier Spectrum Chemical Mfg Cor Supplier No. S1303500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1303, 1310-73-2, NaOH

Spectrum™ Chemical Sodium Hydroxide, Pellets, FCC is used in the chemical washing and peeling of fruits and vegetables and as a thickening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95 to 100.5%
Grade FCC
Packaging Poly Bottle