missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Hydroxide, Pellets, Calbiochem™,

Catalog No. 567530250GM
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass Bottle
Metal Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
567530250GM 250 g Metal Can
567530500GM 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 567530250GM Supplier MilliporeSigma™ Supplier No. 567530250GM

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym NaOH
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color White
Quantity 250 g
Percent Purity ≥95% (Titration)
Packaging Metal Can