missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide (Pellets), ACS Reagent Grade, Ricca Chemical

Catalog No. p-3252320
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
2.5 kg
12 kg
Packaging:
Fiber Drum
Natural Poly Bottle
White Poly Wide Mouth Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCS0530125 125 g White Poly Wide Mouth Bottle
RDCS0530500 500 g White Poly Wide Mouth Bottle
RDCS0530100 1 kg White Poly Wide Mouth Bottle
RDCS053025 2.5 kg Natural Poly Bottle
RDCS053012F 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RDCS0530125 Supplier Ricca Chemical Company Supplier No. RDCS0530125B1
Only null left

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Quantity 125 g
Percent Purity Min. 97.0%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Physical Form Deliquescent Pellets or Flakes
Chemical Name or Material Sodium Hydroxide, Pellets