missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, MP Biomedicals

Supplier:  MP Biomedicals Inc 021534955

 View more versions of this product

Catalog No. ICN1534955


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide
1310-73-2
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Anhydrous
HNaO
500 g
[OH-].[Na+]
39.997
CHEBI:32145

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions