missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, MP Biomedicals
Supplier: MP Biomedicals Inc 0215349501
Specifications
| Sodium Hydroxide | |
| 1310-73-2 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40 |
| Anhydrous | |
| HNaO | |
| 1 kg | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 |
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Safety and Handling
Recommended Storage : Room Temperature