missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide (Pellets/NF/FCC/EP/BP/JP), Fisher Chemical™

Catalog No. p-325943
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S392212 2.5 kg Poly Bottle
S39212 12 kg Poly Pail
S39250 50 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S392212 Supplier Thermo Fisher Scientific Supplier No. S392212
Only null left

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
UN Number 1823
Density 2.13g/cm³
Quantity 2.5 kg
Formula Weight 40
Percent Purity 97.0 to 100.5% (BP)
Grade BP/EP/FCC/JP/NF
Identification Pass Test (BP), Pass Test (BP/EP/FCC/JP/NF), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (NF)
Packaging Poly Bottle
Residual Solvents Meets Requirements (BP), Meets Requirements (BP/EP/NF), Meets Requirements (EP), Meets Requirements (NF)
Vapor Pressure 1mmHg at 739°C
Physical Form Solid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8 : Corrosive