missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide concentrate, For 1 L standard solution, 0.5 M NaOH (0.5 N), Solstice

Catalog No. 6001462
Change view
Click to view available options
Quantity:
6 x 1 Ea.
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001462 1 L
60046707 6 x 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001462 Supplier Solstice Supplier No. 382171EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for 1 L standard solution, 0.5 M NaOH (0.5 N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number UN1824
Quantity 1 L
Formula Weight 40g/mol
Packaging Ampoule
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide concentrate
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338