Learn More
Sodium Hydroxide, Certified, 17.5% (w/w) ±0.5% (w/w), 5.24N (5.24M), LabChem™
Supplier: LabChem LC240782
Specifications
| Sodium Hydroxide | |
| 1310-73-2,7732-18-5 | |
| HNaO | |
| 5.24N (5.24M) | |
| 1.19g/mL | |
| Soluble in water | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40 | |
| Certified | |
| Poly Bottle | |
| Traceable to NIST |
| Colorless | |
| 82.5,17.5 | |
| NaOH | |
| UN1824 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| 1 L | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| 17.5% ±0.5% (w/w) | |
| Passes Test | |
| 1.19g/mL | |
| Liquid |
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature