missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified, 0.5000N ±0.0030N (0.5M), LabChem™

Supplier:  LabChem LC243304

 View more versions of this product

Catalog No. LC243304


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide, 0.5N (0.5M)
1310-73-2,7732-18-5
HNaO
0.5000N ± 0.0030N
1.02g/mL
Soluble in water
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Passes Test
1.02g/mL
Liquid
Colorless
98,2
NaOH
UN1824
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
4 L
[OH-].[Na+]
39.997
CHEBI:32145
Certified
Poly Bottle
Traceable to NIST

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions