missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified, 0.0200N ±0.0002N (0.02M), LabChem™

Supplier:  LabChem LC242204

 View more versions of this product

Catalog No. LC242204


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide
1310-73-2,7732-18-5
HNaO
0.0200N ±0.0002N (0.02M)
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
4 L
[OH-].[Na+]
39.997
CHEBI:32145
Certified
Poly Bottle
Traceable to NIST
Colorless
99.92,0.08
NaOH
1g/mL
Soluble in water
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Passes Test
1g/mL
Liquid

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions