missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Certified, 0.0100N ±0.0001N (0.01M), LabChem™
Supplier: LabChem LC242004
Specifications
| Sodium Hydroxide | |
| 1310-73-2,7732-18-5 | |
| HNaO | |
| 0.0100N ±0.0001N (0.01M) | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| 4 L | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| Certified | |
| Poly Bottle | |
| Traceable to NIST |
| Colorless | |
| 99.96,0.04 | |
| NaOH | |
| 1g/mL | |
| Soluble in water | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40 | |
| Passes Test | |
| 1g/mL | |
| Liquid |
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature