missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Certified, 0.0100N ±0.0001N (0.01M), LabChem™
Supplier: LabChem LC242002
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Sodium Hydroxide | |
1310-73-2,7732-18-5 | |
HNaO | |
0.0100N ±0.0001N (0.01M) | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
1 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Certified | |
Poly Bottle | |
Traceable to NIST |
Colorless | |
99.96,0.04 | |
NaOH | |
1g/mL | |
Soluble in water | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
Passes Test | |
1g/mL | |
Liquid |
Chemical Identifiers
1310-73-2 | |
39.997 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, causticShow More | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature