missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, beads/pellets, 99.99% (metals basis)

CAS: 1310-73-2

$56.14 - $224.19

Chemical Identifiers

CAS 1310-73-2
MDL Number MFCD00003548
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4578006
View Documents
Thermo Scientific Chemicals
4578006
5 g
Each for $56.14
Only null left
 
AA4578014
View Documents
Thermo Scientific Chemicals
04578014
25 g
Each for $102.74
Only null left
 
AA4578022
View Documents
Thermo Scientific Chemicals
04578022
100 g
Each for $224.19
Only null left
 
Description

Description

Sodium hydroxide is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1310-73-2
MFCD00003548

Specifications

White
316°C
2.13
5 g
Odorless
Sodium hydroxide
1310-73-2
MFCD00003548
Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether.
40
3mm Hg at 800°C
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 215-185-5

RTECSNumber : WB4900000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only