missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, ACS Reagent (Pellets), Reagents

Catalog No. p-200021246
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337729 10 kg
01337728 1 kg
01337727 3 kg
01337726 500 g
01337725 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337729 Supplier Labchem Inc Supplier No. C131800010KG
Only null left

Sodium Hydroxide, ACS Reagent (Pellets), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym Lye, Caustic soda
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
Quantity 10 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 2.13
Physical Form Solid
Chemical Name or Material Sodium Hydroxide, ACS Reagent (Pellets)