missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, ACS Reagent (Pellets), Reagents

Catalog No. 01337726
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337726 500 g
01337729 10 kg
01337728 1 kg
01337727 3 kg
01337725 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337726 Supplier Labchem Inc Supplier No. C1318000500A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Hydroxide, ACS Reagent (Pellets), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym Lye, Caustic soda
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.13
Physical Form Solid
Chemical Name or Material Sodium Hydroxide, ACS Reagent (Pellets)