missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071895
Change view
Click to view available options
Quantity:
500 g
6 x 500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001417 500 g
60045313 6 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001417 Supplier Honeywell Chemicals Supplier No. 221465500G
Only null left

ACS Reagent, pellets,≥97.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number 1823
Solubility Information Soluble in water, 1260 g/L at 20°C
Quantity 500 g
Formula Weight 40g/mol
Percent Purity ≥97%
Grade ACS Reagent
Vapor Density >1 (vs air)
Vapor Pressure 18mmHg (20°C), 3mmHg (37°C)
Chemical Name or Material Sodium Hydroxide
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H314
EINECSNumber 215-185-5
RTECSNumber WB4900000