missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 98%, pure, flakes

Catalog No. p-4813648
missing translation for 'changeView'
Click to view available options
Quantity:
2.5 kg
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450580025 2.5 kg
AC450580250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AC450580025 Supplier Thermo Scientific Chemicals missing translation for 'supplierNo' 450580025
Only null left

CAS: 1310-73-2 | HNaO | 39.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 1390.0°C
Melting Point 318.0°C
Assay Percent Range 98%
Linear Formula NaOH
Solubility Information Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone
Quantity 2.5 kg
Formula Weight 40
Percent Purity 98%
Grade Pure
Packaging Plastic Bottle
Chemical Name or Material Sodium Hydroxide
Show More Show Less

RUO – Research Use Only