missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 97+%, ACS reagent, pellets

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Supplier:  Thermo Scientific Chemicals 424330100

 View more versions of this product

Catalog No. AC424330100

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium hydroxide
1390.0°C
318.0°C
HNaO
01,1083; 05,616; 07,336; 08,460
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
10 kg
[OH-].[Na+]
39.997
CHEBI:32145
≥97%
Plastic drum
Pellet
White
1310-73-2
97+%
NaOH
15, 8761
Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
ACS Reagent
1mbar at 700°C

Chemical Identifiers

1310-73-2
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.

Emergency Overview
Causes severe burns by all exposure routes. Hygroscopic.
NFPA
Health: 3
Flammability:

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only