missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, ≥97.0% (T), Honeywell™
Purum p.a.,≥97.0% (T), pellets
$1135.08 - $1135.08
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| MDL Number | MFCD00003548 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
Specifications
| 1310-73-2 | |
| NaOH | |
| UN1823 | |
| 5 kg | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| ≥97.0% (T) | |
| >1 (vs air) | |
| Sodium hydroxide |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40g/mol | |
| Poly Bottle | |
| 18mmHg (20°C), 3mmHg (37°C) |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation : 8 / PGII
RTECSNumber : 215-185-5