missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 96+% Caustic Soda Beads, Reagents

Catalog No. 01337015
Change view
Click to view available options
Quantity:
500 g
3 kg
10 lb.
10 kg
50 lb.
50 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337015 10 kg
01337016 10 lb.
01337013 3 kg
01337012 500 g
01337011 50 kg
01337010 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01337015 Supplier Labchem Inc Supplier No. C231825010KG
Only null left

Sodium Hydroxide, 96+% Caustic Soda Beads, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym Lye, Caustic soda
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
Quantity 10 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 2.13
Physical Form Solid
Chemical Name or Material Sodium Hydroxide, 96+% Caustic Soda Beads