missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 96+% Caustic Soda Beads, Reagents

Sodium Hydroxide, 96+% Caustic Soda Beads, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc C231825025LB

 View more versions of this product

Catalog No. 01-337-014

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

14
White
1310-73-2
HNaO
25 lb.
[OH-].[Na+]
40.00
Fiber Drum
Solid
Sodium Hydroxide, 96+% Caustic Soda Beads
1390°C
318°C
Lye, Caustic soda
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
Pure
2.13

Chemical Identifiers

1310-73-2
40.00
Lye, Caustic soda
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions