missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 96+% Caustic Soda Beads, Reagents
Specifications
Specifications
| pH | 14 |
| Color | White |
| Boiling Point | 1390°C |
| Melting Point | 318°C |
| Quantity | 50 kg |
| Grade | Pure |
| Packaging | Fiber Drum |
| Specific Gravity | 2.13 |
| Physical Form | Solid |
| Chemical Name or Material | Sodium Hydroxide, 96+% Caustic Soda Beads |