missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% w/w Reagent (760 g/L), Reagents
Sodium Hydroxide, 50% w/w Reagent (760 g/L), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1318600200E*
Specifications
| >13 | |
| Colorless | |
| 7732-18-5,1310-73-2 | |
| H2O | |
| 200 L | |
| O | |
| 18.02 | |
| Poly Drum | |
| Liquid |
| Sodium Hydroxide, 50% w/w Reagent (760 g/L) | |
| 140°C | |
| 12°C | |
| Lye, Caustic soda | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.5267 |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Lye, Caustic soda | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |