missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% w/w Reagent (760 g/L), Reagents

Sodium Hydroxide, 50% w/w Reagent (760 g/L), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc C1318600500A

 View more versions of this product

Catalog No. 01-337-719

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

>13
Colorless
7732-18-5,1310-73-2
H2O
500 mL
O
18.02
Poly Bottle
Liquid
Sodium Hydroxide, 50% w/w Reagent (760 g/L)
140°C
12°C
Lye, Caustic soda
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.5267

Chemical Identifiers

7732-18-5
18.02
Lye, Caustic soda
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions