missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% (w/v) Solution, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S2241LTPL
Specifications
| Sodium Hydroxide Solution | |
| 1 L | |
| Chemical Solution |
| 1310-73-2,7732-18-5 | |
| 50% ±1% | |
| Poly Bottle |
Chemical Identifiers
| 1310-73-2 |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.