missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 40% (w/w), Analytical Reagent Grade, Ricca Chemical
For Kjeldahl Nitrogen analysis
Supplier: Ricca Chemical Company 728825
Specifications
| Sodium Hydroxide | |
| 1310-73-2,7732-18-5 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium hydroxide | |
| 14798 | |
| 39.8 to 40.2% | |
| 40% (w/w) Aqueous Solution |
| Nitrogen Free | |
| HNaO | |
| 10 L | |
| [OH-].[Na+] | |
| 40.00 | |
| CHEBI:32145 | |
| Analytical | |
| Cubitainer |
Chemical Identifiers
| 1310-73-2 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| [OH-].[Na+] |
| HNaO | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 |