missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 40% (w/v), Nitrogen Free - Suitable for Kjeldahl Nitrogen Analysis, Ricca Chemical

Supplier:  Ricca Chemical Company 72805

 View more versions of this product

Catalog No. 7280-5

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide Solution
40% (w/v), Nitrogen Free
HNaO
20 L
[OH-].[Na+]
40.00
CHEBI:32145
Laboratory
Cubitainer
Liquid
Colorless
1310-73-2,7732-18-5
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
14798
39.8 to 40.2%
40% (w/v)
Aqueous Solution

Chemical Identifiers

1310-73-2
HEMHJVSKTPXQMS-UHFFFAOYSA-M
CHEBI:32145
HNaO
14798
[OH-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 36 months

SDS Documents

Documents

Promotions

Promotions