missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 40% (w/v), Nitrogen Free - Suitable for Kjeldahl Nitrogen Analysis, Ricca Chemical
Supplier: Ricca Chemical Company 72801
Specifications
| Sodium Hydroxide Solution | |
| 40% (w/v), Nitrogen Free | |
| HNaO | |
| 4 L | |
| [OH-].[Na+] | |
| 40.00 | |
| CHEBI:32145 | |
| Laboratory | |
| Natural Poly Bottle | |
| Liquid |
| Colorless | |
| 1310-73-2,7732-18-5 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium hydroxide | |
| 14798 | |
| 39.8 to 40.2% | |
| 40% (w/v) | |
| Aqueous Solution |
Chemical Identifiers
| 1310-73-2 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| CHEBI:32145 |
| HNaO | |
| 14798 | |
| [OH-].[Na+] |
Safety and Handling
ShelfLife : 36 months